Learn More
Medchemexpress LLC HY-50682 10mg Medchemexpress, Azeliragon CAS:603148-36-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY5068210MG
Encompass_Preferred
Medchemexpress, HY-50682 10mg Azeliragon CAS:603148-36-3 Azeliragon (TTP488) is an orally bioavailable inhibitor of the receptor for advanced glycation end products (RAGE) in development as a potential treatment to slow disease progression in patients with mild Alzheimer’s disease (AD). Azeliragon also can cross the blood-brain barrier (BBB)[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9327