Learn More
Cayman Chemical Azelnidipine, Purity: ≥98%, CAS: 123524-52-7, 1 g
SDP
Catalog No. 502985054
Encompass_Preferred
A dihydropyridine L-type calcium channel blocker with a slow onset profile that displays antihypertensive activity
Catalog No. 50-298-5054 Supplier Cayman Chemical Supplier No. 212491 G
A dihydropyridine L-type calcium channel blocker with a slow onset profile that displays antihypertensive activity