Learn More
Medchemexpress LLC Azemiglitazone potas 50mg | 1314533-27-1 | 50MG
SDP
Catalog No. 5000243012
Encompass_Preferred
Azemiglitazone potassium (MSDC-0602K) a PPAR -sparing thiazolidinedione (Ps-TZD) binds to PPAR with the IC50 of 18 25 M[1 Azemiglitazone potassium modulates the mitochondrial pyruvate carrier (MPC) Azemiglitazone potassium can be used for the research of fatty liver including dysfunctional lipid metabolism inflammation and insulin resistance[2 Azemiglitazone potassium an insulin sensitizer improves insulinemia and fatty liver disease in mice alone and in combination with Liraglutide[3
Catalog No. 50-002-43012 Supplier Medchemexpress LLC Supplier No. HY108022A50MG
Azemiglitazone potassium (MSDC-0602K) a PPAR -sparing thiazolidinedione (Ps-TZD) binds to PPAR with the IC50 of 18 25 M[1 Azemiglitazone potassium modulates the mitochondrial pyruvate carrier (MPC) Azemiglitazone potassium can be used for the research of fatty liver including dysfunctional lipid metabolism inflammation and insulin resistance[2 Azemiglitazone potassium an insulin sensitizer improves insulinemia and fatty liver disease in mice alone and in combination with Liraglutide[3