Learn More
Medchemexpress LLC Azemiglitazone potassium | 1314533-27-1 | 99.5% | 409.50 | C19H16KNO5S | 10 MG
SDP
Catalog No. 5000243082
Encompass_Preferred
Azemiglitazone potassium is the potassium salt of azemiglitazone, a thiazolidinedione-like insulin sensitizer that modulates the mitochondrial pyruvate carrier (MPC) with low PPARγ activation. It is provided as a high-purity research compound for in vitro and preclinical studies of metabolic disorders such as diabetes and nonalcoholic steatohepatitis (NASH).
  • High purity suitable for research applications.
  • Potassium salt form enhances handling and solubility in polar solvents.
  • High DMSO solubility (250 mg/mL) with ultrasonic assistance.
  • Recommended sealed storage away from moisture; solvent storage guidelines apply.
  • Modulates mitochondrial pyruvate carrier and acts as an insulin sensitizer for metabolic research.
Catalog No. 50-002-43082 Supplier Medchemexpress LLC Supplier No. HY108022A10MG
Azemiglitazone potassium is the potassium salt of azemiglitazone, a thiazolidinedione-like insulin sensitizer that modulates the mitochondrial pyruvate carrier (MPC) with low PPARγ activation. It is provided as a high-purity research compound for in vitro and preclinical studies of metabolic disorders such as diabetes and nonalcoholic steatohepatitis (NASH).
  • High purity suitable for research applications.
  • Potassium salt form enhances handling and solubility in polar solvents.
  • High DMSO solubility (250 mg/mL) with ultrasonic assistance.
  • Recommended sealed storage away from moisture; solvent storage guidelines apply.
  • Modulates mitochondrial pyruvate carrier and acts as an insulin sensitizer for metabolic research.