Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Azemiglitazone potassium | 1314533-27-1 | 99.5% | 409.50 | C19H16KNO5S | 25 MG
SDP

Supplier:  Medchemexpress LLC HY108022A25MG

Encompass_Preferred

Azemiglitazone potassium is the potassium salt form of azemiglitazone, a thiazolidinedione-related research compound used as an insulin sensitizer and mitochondrial pyruvate carrier modulator. Supplied as a high-purity white to off-white solid for laboratory research use only.

  • Purity 99.5%.
  • Potassium salt form suitable for salt-specific studies.
  • White to off-white solid, convenient for handling and weighing.
  • Molecular weight 409.50; formula C19H16KNO5S.
  • Soluble in DMSO at 250 mg/mL; ultrasonic assistance may be required.
  • Intended for research use only; not for human or clinical use.
  • Supplied in 25 MG quantity.

Catalog No. 50-002-43084


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.