Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Azemiglitazone potassium | 1314533-27-1 | 99.5% | 409.50 g/mol | C19H16KNO5S | 5 MG
SDP

Catalog No. 5000243096
Encompass_Preferred

Azemiglitazone potassium is the potassium salt of azemiglitazone, a thiazolidinedione-like insulin sensitizer used in preclinical research on metabolic disorders such as type 2 diabetes and non-alcoholic steatohepatitis. Supplied as a solid with high analytical purity and accompanied by datasheet, certificate of analysis, and safety data sheet for research use.

  • High purity for reliable analytical and pharmacological studies.
  • Molecular weight 409.50 g/mol.
  • Chemical formula C19H16KNO5S.
  • Available in small milligram pack sizes suitable for laboratory experiments.
  • Supplied with datasheet, COA, and SDS for documentation and handling guidance.

Catalog No. 50-002-43096 Supplier Medchemexpress LLC Supplier No. HY108022A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Azemiglitazone potassium is the potassium salt of azemiglitazone, a thiazolidinedione-like insulin sensitizer used in preclinical research on metabolic disorders such as type 2 diabetes and non-alcoholic steatohepatitis. Supplied as a solid with high analytical purity and accompanied by datasheet, certificate of analysis, and safety data sheet for research use.

  • High purity for reliable analytical and pharmacological studies.
  • Molecular weight 409.50 g/mol.
  • Chemical formula C19H16KNO5S.
  • Available in small milligram pack sizes suitable for laboratory experiments.
  • Supplied with datasheet, COA, and SDS for documentation and handling guidance.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.