Learn More
Medchemexpress LLC Azepan-2-one-d10 | 169297-53-4 | 99.8% | 123.22 | 100 MG
SDP
Catalog No. 5000453561
Encompass_Preferred
Azepan-2-one-d10 is a deuterium labeled compound designed for research applications. It serves as a tracer and an internal standard for quantitative analysis using techniques like NMR, GC-MS, or LC-MS. Deuteration of drug molecules can influence their pharmacokinetic and metabolic profiles, making this compound valuable for drug development studies.
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Facilitates studies on the effects of deuteration on pharmacokinetic and metabolic profiles of drugs
  • Incorporates stable heavy isotopes for quantitation during drug development
Catalog No. 50-004-53561 Supplier Medchemexpress LLC Supplier No. HYW010542S100MG
Azepan-2-one-d10 is a deuterium labeled compound designed for research applications. It serves as a tracer and an internal standard for quantitative analysis using techniques like NMR, GC-MS, or LC-MS. Deuteration of drug molecules can influence their pharmacokinetic and metabolic profiles, making this compound valuable for drug development studies.
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Facilitates studies on the effects of deuteration on pharmacokinetic and metabolic profiles of drugs
  • Incorporates stable heavy isotopes for quantitation during drug development