Learn More
Medchemexpress LLC Azepan-2-one-d10 | 169297-53-4 | 99.8% | 123.22 | 5 MG
SDP
Catalog No. 5000451584
Encompass_Preferred
Azepan-2-one-d10 is the deuterium labeled Azepan-2-one. This compound is intended for research use only. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes have been incorporated into drug molecules as tracers for quantitation.
  • Deuteration has potential to impact the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-51584 Supplier Medchemexpress LLC Supplier No. HYW010542S5MG
Azepan-2-one-d10 is the deuterium labeled Azepan-2-one. This compound is intended for research use only. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes have been incorporated into drug molecules as tracers for quantitation.
  • Deuteration has potential to impact the pharmacokinetic and metabolic profiles of drugs.