Learn More
Medchemexpress LLC Azepan-2-one-d10 | 169297-53-4 | 99.8% | 123.22 | 50 MG
SDP
Catalog No. 5000452917
Encompass_Preferred
Azepan-2-one-d10 is the deuterium labeled Azepan-2-one. This product is for research use only and not intended for sale to patients. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules primarily as tracers for quantitation during the drug development process. Deuteration has garnered attention due to its potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be utilized as a tracer.
  • Can serve as an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS.
Catalog No. 50-004-52917 Supplier Medchemexpress LLC Supplier No. HYW010542S50MG
Azepan-2-one-d10 is the deuterium labeled Azepan-2-one. This product is for research use only and not intended for sale to patients. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules primarily as tracers for quantitation during the drug development process. Deuteration has garnered attention due to its potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be utilized as a tracer.
  • Can serve as an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS.