Learn More
Medchemexpress LLC Azepexole (hydrochloride) | 147663-20-5 | MFCD35200177 | >98.0% | 217.7 g/mol | C9H16ClN3O | 5 MG
SDP
Catalog No. 5000247593
Encompass_Preferred
Azepexole hydrochloride is the hydrochloride salt of azepexole (B-HT 933), a selective α2-adrenoceptor agonist used as a research reagent to study adrenergic receptor pharmacology and related physiological effects such as analgesia and antitussive responses.
  • Selective α2-adrenoceptor agonist used in receptor pharmacology studies.
  • Reported purity greater than 98% (HPLC).
  • Molecular formula C9H16ClN3O and molecular weight 217.7 g/mol.
  • Provided as a small-scale research pack suitable for laboratory experiments.
  • Intended for research use only; not for human or clinical use.
Catalog No. 50-002-47593 Supplier Medchemexpress LLC Supplier No. HY103212A5MG
Azepexole hydrochloride is the hydrochloride salt of azepexole (B-HT 933), a selective α2-adrenoceptor agonist used as a research reagent to study adrenergic receptor pharmacology and related physiological effects such as analgesia and antitussive responses.
  • Selective α2-adrenoceptor agonist used in receptor pharmacology studies.
  • Reported purity greater than 98% (HPLC).
  • Molecular formula C9H16ClN3O and molecular weight 217.7 g/mol.
  • Provided as a small-scale research pack suitable for laboratory experiments.
  • Intended for research use only; not for human or clinical use.