Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Azer Scientific 12-Place Prostate Biopsy Kit
SDP

Catalog No. 22026202
Change view
Click to view available options
Capacity (Metric):
20 mL, pre-filled 10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
22-026-202 20 mL, pre-filled 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-026-202 Supplier Azer Scientific Supplier No. 10820
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ready to use

  • Features twelve 20mL vials filled with 10mL of 10% neutral buffered formalin
  • Vials are 95kPa compliant with a leak-resistant seal
  • Each kit includes specific instructions with lot numbers and expiration dates of the formalin product
  • All specimen locations are clearly marked within the kits

Specifications

Capacity (Metric) 20 mL, pre-filled 10 mL
Includes Specific instructions with lot numbers and expiration dates of the formalin product
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.