Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Azer Scientific Formagel™ Tissue Fixation and Transport Gel
SDP

Catalog No. 22026355
Change view
Click to view available options
Quantity:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-026-355 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-026-355 Supplier Azer Scientific Supplier No. PFG20
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Solution to leaking formalin problems for improved histology studies

  • Maintains specimen integrity
  • Fixation results identical to standard 10% Neutral Buffered Formalin
  • Viscoscity enables safe transport without spills

For leak-free transport and fixation of tissue specimens. Compatible with routine, special and IHC staining procedures.

Specifications

Closure Type Screw-on
Color Clear
Content And Storage 10% Neutral Buffered Formalin
Quantity 10mL
For Use With (Application) Routine, special, IHC staining procedures
Material Polypropylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.