Learn More
Azer Scientific Formalock™ Pre-Filled Formalin Wide Containers
SDP
Catalog No. 22026502
Change view
Click to view available options
No. per Pack:
72/Cs.
Catalog No. No. per Pack
22-026-502 72/Cs.
1 options
This item is not returnable. View return policy
Catalog No. 22-026-502 Supplier Azer Scientific Supplier No. BPFNBF60W
This item is not returnable. View return policy
Eliminates annoyance and hazard of leaking prefills
  • Patented design
  • Audible “click« when achieving full seal closure
  • Designed with wide mouth opening, allowing for larger specimens with less waste
  • Leak tested
  • Hospital laboratories
  • Reference laboratories
  • Physician's office laboratories - GI, Urology & Dermatology
  • Education/University Pathology Departments
Specifications
No. per Pack 72/Cs.