Learn More
Azer Scientific Glucose Tolerance Beverages
SDP
Catalog No. 22026203
Change view
Click to view available options
Flavor:
Fruit Punch
Lemon-Lime
Orange
Quantity:
50 g
75 g
100 g
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Flavor Quantity
22-026-203 Orange 50 g
22-026-204 Orange 75 g
22-026-205 Orange 100 g
22-026-208 Fruit Punch 100 g
22-026-207 Fruit Punch 75 g
22-026-206 Fruit Punch 50 g
22-026-209 Lemon-Lime 50 g
22-026-210 Lemon-Lime 75 g
22-026-211 Lemon-Lime 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Catalog No. 22-026-203 Supplier Azer Scientific Supplier No. 10O050
 Restricted Item
Add to Cart
Add to Cart
This item is not returnable. View return policy
Available in lemon lime, fruit punch, and orange flavors
  • Produced in an FDA certified pharmaceutical manufacturing facility
  • Dairy free, gluten free, and certified kosher product
  • Shatterproof plastic bottles
  • Manufactured according to WHO and ADA standards
  • Expiration date on product determined via rigorous accelerated and real time stability studies
  • Integrity of every batch certified by an FDA recognized independent laboratory
  • All flavors are available in standard 50, 75 and 100 gram concentrations
Specifications
Certifications/Compliance FDA, WHO, ADA
Content And Storage Room Temperature
Flavor Orange
Quantity 50 g