Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Azer Scientific Rapid-Freeze™ Rapid Freezing Spray
SDP

Catalog No. 22026200
Change view
Click to view available options
Product Type:
Rapid-Freeze Spray
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
22-026-200 Rapid-Freeze Spray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-026-200 Supplier Azer Scientific Supplier No. RAPFRZ
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Instantly cool or freeze routine histology samples and tissue blocks.

  • Nonflammable, moisture-free spray used to instantly cool or freeze samples for routine histology
  • Brings samples to -60°C in under three seconds
  • Contains no CFCs or HFCs so it will not deplete stratospheric ozone
  • Proprietary Rapid Zap™ device provides a more precise distribution of freeze spray versus standard applicator

Specifications

Applications Instantly cool or freeze routine histology samples and tissue blocks
Quantity 12 x 10 oz.
Product Type Rapid-Freeze Spray
Packaging Can
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.