Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Azetidine

Catalog No. 281069250MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
28-106-9250MG 250 mg
28-106-91G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 28-106-9250MG Supplier Sigma Aldrich Supplier No. 281069250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Phototransformations of azetidine radical cations in freonic matrices under the action of light with ++ = 436nm has been investigated. the IR spectrum of azetidine in solid argon matrices has been measured.azetidine was employed in:
  • A high yielding palladium-catalyzed cross-coupling raection with aryl bromides
  • Ullmann type coupling reaction with iodonitroflourenes

Specifications

CAS 503-29-7
Density 0.847 g/mL (at 25°C (literature))
Boiling Point 61°C to 62°C (lit.)
Molecular Formula C3H7N
Refractive Index n20/D 1.432 (literature)
Linear Formula C3H7N
MDL Number MFCD00005165
Quantity 250 mg
Synonym Trimethylene imine
Molecular Weight (g/mol) 57.09
Percent Purity 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.