Learn More
Medchemexpress LLC Azetidine-2-carboxylic acid | 2517-04-6 | 98.0% | C4H7NO2 | 1 ML
SDP
Catalog No. 5000434629
Encompass_Preferred
Azetidine-2-carboxylic acid is a non-proteinogenic amino acid homologue of proline, commonly found in beets. It can be misincorporated into proteins in place of proline in many species, including humans. This compound is described as a toxic and teratogenic agent, and has been shown to incorporate into collagen synthesized by chick embryo cartilage in vitro.
  • Non-proteinogenic amino acid homologue of proline
  • Found in common beets
  • Can be misincorporated into proteins in place of proline in many species
  • Considered a toxic and teratogenic agent
  • Incorporated into collagen synthesized by chick embryo cartilage in vitro
  • Solid, white to off-white appearance
Catalog No. 50-004-34629 Supplier Medchemexpress LLC Supplier No. HY7530810MM/1ML
Azetidine-2-carboxylic acid is a non-proteinogenic amino acid homologue of proline, commonly found in beets. It can be misincorporated into proteins in place of proline in many species, including humans. This compound is described as a toxic and teratogenic agent, and has been shown to incorporate into collagen synthesized by chick embryo cartilage in vitro.
  • Non-proteinogenic amino acid homologue of proline
  • Found in common beets
  • Can be misincorporated into proteins in place of proline in many species
  • Considered a toxic and teratogenic agent
  • Incorporated into collagen synthesized by chick embryo cartilage in vitro
  • Solid, white to off-white appearance