Learn More
Medchemexpress LLC Azetidine-2-carboxylic acid | 2517-04-6 | 98.0% | C4H7NO2 | 25 G
SDP
Catalog No. 5000357608
Encompass_Preferred
Azetidine-2-carboxylic acid is a non-proteinogenic amino acid homologue of proline, found in common beets. It can be misincorporated into proteins in place of proline in many species, including humans, and is considered a toxic and teratogenic agent. This compound is intended for research use only.
  • Non-proteinogenic amino acid homologue of proline
  • Found in common beets
  • White to off-white solid appearance
  • For research use only
  • Store solid at 4°C, sealed, away from moisture
  • Store in solvent at -80°C for 6 months or -20°C for 1 month
  • Soluble in water (50 mg/mL)
Catalog No. 50-003-57608 Supplier Medchemexpress LLC Supplier No. 25GHY75308
Azetidine-2-carboxylic acid is a non-proteinogenic amino acid homologue of proline, found in common beets. It can be misincorporated into proteins in place of proline in many species, including humans, and is considered a toxic and teratogenic agent. This compound is intended for research use only.
  • Non-proteinogenic amino acid homologue of proline
  • Found in common beets
  • White to off-white solid appearance
  • For research use only
  • Store solid at 4°C, sealed, away from moisture
  • Store in solvent at -80°C for 6 months or -20°C for 1 month
  • Soluble in water (50 mg/mL)