Learn More
Medchemexpress LLC Azetidine-2-carboxylic acid | 2517-04-6 | 98.0% | C4H7NO2 | 5 G
SDP
Catalog No. 5000434394
Encompass_Preferred
Azetidine-2-carboxylic acid is a non-proteinogenic amino acid homologue of proline. It is found in common beets and can be misincorporated into proteins in place of proline in many species, including humans. It is described as a toxic and teratogenic agent.
  • Non-proteinogenic amino acid homologue of proline
  • Found in common beets
  • Can be misincorporated into proteins
  • Toxic and teratogenic agent
  • Incorporated into collagen synthesized by chick embryo cartilage in vitro
Catalog No. 50-004-34394 Supplier Medchemexpress LLC Supplier No. HY753085G
Azetidine-2-carboxylic acid is a non-proteinogenic amino acid homologue of proline. It is found in common beets and can be misincorporated into proteins in place of proline in many species, including humans. It is described as a toxic and teratogenic agent.
  • Non-proteinogenic amino acid homologue of proline
  • Found in common beets
  • Can be misincorporated into proteins
  • Toxic and teratogenic agent
  • Incorporated into collagen synthesized by chick embryo cartilage in vitro