Learn More
Selleck Chemical LLC Azetidine-2-carboxylic acid
SDP
Supplier:  Selleck Chemical LLC S557225MG
Encompass_Preferred
Azetidine-2-carboxylic acid (L-Aze) is a toxic and teratogenic non-protein amino acid that is misincorporated into protein in place of proline altering collagen keratin hemoglobin and protein folding
Catalog No. 50-000-03136