Learn More
Selleck Chemical LLC Azetidine-2-carboxylic acid
SDP
Catalog No. 5000003818
Encompass_Preferred
Azetidine-2-carboxylic acid (L-Aze) is a toxic and teratogenic non-protein amino acid that is misincorporated into protein in place of proline altering collagen keratin hemoglobin and protein folding
Catalog No. 50-000-03818 Supplier Selleck Chemical LLC Supplier No. S55721G
Azetidine-2-carboxylic acid (L-Aze) is a toxic and teratogenic non-protein amino acid that is misincorporated into protein in place of proline altering collagen keratin hemoglobin and protein folding