Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Azetidine-3-carboxylic acid | 36476-78-5 | MFCD00191763 | 101.10 | C4H7NO2 | 10 G
SDP

Catalog No. 5000203887
Encompass_Preferred

Azetidine-3-carboxylic acid is a small heterocyclic carboxylic acid used as a building block in medicinal and linker chemistry, including applications in ADC and PROTAC synthesis.

  • Small four-membered heterocyclic carboxylic acid building block.
  • Used in linker and medicinal chemistry, including ADC and PROTAC synthesis.
  • Physical form: powder suitable for storage under recommended conditions.
  • Molecular formula C4H7NO2 and molecular weight 101.10.
  • Storage stability: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-03887 Supplier Medchemexpress LLC Supplier No. HYY053010G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Azetidine-3-carboxylic acid is a small heterocyclic carboxylic acid used as a building block in medicinal and linker chemistry, including applications in ADC and PROTAC synthesis.

  • Small four-membered heterocyclic carboxylic acid building block.
  • Used in linker and medicinal chemistry, including ADC and PROTAC synthesis.
  • Physical form: powder suitable for storage under recommended conditions.
  • Molecular formula C4H7NO2 and molecular weight 101.10.
  • Storage stability: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.