Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Azetidine (hydrochloride) | 36520-39-5 | 97.0% | 93.56 | 10 G
SDP

Supplier:  Medchemexpress LLC HYW01062810G

Encompass_Preferred

Azetidine (Trimethyleneimine) hydrochloride is a biochemical reagent that can be used as a biological material or organic compound for life science-related research. It is also a nitrogen-containing heterocyclic compound, which can be used in the synthesis of other active compounds.

  • Can be used as a biochemical reagent
  • Suitable for life science-related research
  • Nitrogen-containing heterocyclic compound
  • Used in the synthesis of other active compounds
  • Formula: C3H8ClN
  • Appearance: Solid
  • Color: White to off-white
  • Shipping conditions: Room temperature (continental US)
  • Storage conditions: 4°C, sealed, away from moisture

Catalog No. 50-004-52925


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.