Learn More
Medchemexpress LLC AzGGK (TFA) | 2407768-11-8 | 99.8% | 400.31 | 5 MG
SDP
Catalog No. 5000361643
Encompass_Preferred
AzGGK (TFA) is an unnatural amino acid site-specifically incorporated into proteins via genetic-code expansion. It functions as a site-specific probe for ubiquitylation and SUMOylation. This compound also serves as a click chemistry reagent, featuring an azide group for copper-catalyzed azide-alkyne cycloaddition (CuAAc) and strain-promoted alkyne-azide cycloaddition (SPAAC) reactions.
  • Unnatural amino acid
  • Site-specifically incorporated into proteins
  • Functions as a site-specific probe for ubiquitylation and SUMOylation
  • Click chemistry reagent
  • Contains an azide group
  • Engages in copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC)
Catalog No. 50-003-61643 Supplier Medchemexpress LLC Supplier No. HY148835A5MG
AzGGK (TFA) is an unnatural amino acid site-specifically incorporated into proteins via genetic-code expansion. It functions as a site-specific probe for ubiquitylation and SUMOylation. This compound also serves as a click chemistry reagent, featuring an azide group for copper-catalyzed azide-alkyne cycloaddition (CuAAc) and strain-promoted alkyne-azide cycloaddition (SPAAC) reactions.
  • Unnatural amino acid
  • Site-specifically incorporated into proteins
  • Functions as a site-specific probe for ubiquitylation and SUMOylation
  • Click chemistry reagent
  • Contains an azide group
  • Engages in copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC)