Learn More
Aviva Systems Biology Corp AZI2 Antibody - middle region (ARP62739_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62739_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against AZI2. It was validated on Western Blot. . Alternate names: AZ2, NAP1, TILPApplicationWB. Species reactivity: Dog, Horse, Human, Pig, Rabbit, Rat
Catalog No. 50-287-761