Learn More
Aviva Systems Biology Corp AZI2 Antibody - N-terminal region (ARP62738_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62738_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against AZI2. It was validated on Western Blot. . Alternate names: AZ2, NAP1, TILPApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-287-760