Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Azide-A-Dsbso Crosslinker

Catalog No. 909629100MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-962-9100MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 90-962-9100MG Supplier Sigma Aldrich Supplier No. 909629100MG
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Azide-A-DSBSO (azide-tagged, acid-cleavable disuccinimidyl bissulfoxide) Crosslinker is a homobifunctional, membrane-permeable, enrichable disulfoxide-containing crosslinker for analysis of protein-protein interactions (PPIs) through crosslinking mass spectrometry (XL-MS). Azide-A-DSBSO possesses two N-hydroxysuccinimide (NHS) ester groups for targeting amines, a ≥14 +a spacer length, two symmetrical acid-cleavable C-S bonds, and a central bioorthogonal azide tag. After reacting with lysine (Lys) residues, the azide tag permits selective enrichment of crosslinked proteins or peptides with a biotin-conjugated alkyne probe to enhance their detection. Additionally, the post-cleavage spacer yields tagged peptides for unambiguous identification by collision-induced dissociation in tandem MS. Azide-A-DSBSO Crosslinker has mapped in vivo PPIs for target protein complexes and whole proteomes fro mLiving cells. It will also be a useful proteomics tool in the quest for targeting undruggable protein targets.Technology Spotlight: Cross-Linkers for Elucidation of Protein-Protein InteractionsA New in Vivo Cross-linking Mass Spectrometry Platform to Define Protein-Protein Interactions in Living CellsSynthesis of two new enrichable and MS-cleavable cross-linkers to define protein-protein interactions by mass spectrometrySubject to US Patent Application #15/275,001 of the Regents of the University of California

Specifications

CAS 2133345-76-1
Molecular Formula C24H29N3O9
Linear Formula C27H18B2ClF8N5Pd
Quantity 50 mg
Synonym 7-(2-(2-((2-(2,6-Dioxopiperidin-3-yl)-1,3-dioxoisoindolin-4-yl)amino)-2-oxoethoxy)ethoxy)heptanoic acid; Crosslinker-E3 ligase ligand conjugate; Pomalidomide conjugate; Pomalidomide-2-2-6-acid; Protein degrader building block for PROTAC(R) research; Template for synthesis of targeted protein degrader
Molecular Weight (g/mol) 503.5 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.