Learn More
Biotium Azide Alkyne BCN Tetrazine TCO Reactive CF Dyes Other Dyes Biotinylation Reagents Reactive CF Dyes CF405 1/EA
Catalog No. 5000501488
Encompass_Preferred
CF405S DBCO (Dibenzocyclooctyne) is a highly efficient fast-reacting alternative to fluorescently labeled BCN and DIBO DBCO-azide reactions do not require toxic copper catalysts (CuAAC) making them more suitable for live-cell or in vivo experiments
Catalog No. 50-005-01488 Supplier Biotium Supplier No. 96111
CF405S DBCO (Dibenzocyclooctyne) is a highly efficient fast-reacting alternative to fluorescently labeled BCN and DIBO DBCO-azide reactions do not require toxic copper catalysts (CuAAC) making them more suitable for live-cell or in vivo experiments