Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LUNA NANOTECH INC Azide Coated Magnetic Beads, 500 Nm, 10 ML

Catalog No. 502566984
Encompass_Preferred

500 nm diameter magnetic beads with Azide (N=N=N) functionalized surface.

Features:
- Conjugation ready: Activated for click chemistry conjugation with DBCO containing molecules.
- High monodispersity.
- Stable in high salt conditions.
- Fast response to a magnet.

Applications:
- Can be used for magnetic separation of DNA, RNA, proteins, antibodies, cells and small molecules from complex samples.
- DBCO-functionalized proteins, small molecules, DNA / RNA can be covalently attached to bead surface through click chemistry conjugation method.

Catalog No. 50-256-6984 Supplier LUNA NANOTECH INC Supplier No. MGBAZD1010
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

500 nm diameter magnetic beads with Azide (N=N=N) functionalized surface.

Features:
- Conjugation ready: Activated for click chemistry conjugation with DBCO containing molecules.
- High monodispersity.
- Stable in high salt conditions.
- Fast response to a magnet.

Applications:
- Can be used for magnetic separation of DNA, RNA, proteins, antibodies, cells and small molecules from complex samples.
- DBCO-functionalized proteins, small molecules, DNA / RNA can be covalently attached to bead surface through click chemistry conjugation method.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.