Learn More
Medchemexpress LLC Azide-PEG-amine | 00-00-0 | 95.0% | N3-PEG-NH2 (polymeric, avg. MW 2000) | 10MG
SDP
Catalog No. 5000258521
Encompass_Preferred
Azide-PEG-amine (average MW 2000) is a bifunctional polyethylene glycol linker with an azide group and a primary amine, supplied as a white to off-white solid. It is intended for click chemistry and conjugation workflows, enabling both copper-catalyzed and strain-promoted azide-alkyne cycloadditions for linker assembly, PEGylation, and bioconjugation applications.
  • Bifunctional azide and amine termini for orthogonal conjugation.
  • Average molecular weight 2000 for flexible spacer properties.
  • High purity (95.0%) suitable for research-grade conjugation reactions.
  • White to off-white solid for easy handling.
  • Stable under recommended storage conditions to preserve activity.
Catalog No. 50-002-58521 Supplier Medchemexpress LLC Supplier No. HY14066110MG
Azide-PEG-amine (average MW 2000) is a bifunctional polyethylene glycol linker with an azide group and a primary amine, supplied as a white to off-white solid. It is intended for click chemistry and conjugation workflows, enabling both copper-catalyzed and strain-promoted azide-alkyne cycloadditions for linker assembly, PEGylation, and bioconjugation applications.
  • Bifunctional azide and amine termini for orthogonal conjugation.
  • Average molecular weight 2000 for flexible spacer properties.
  • High purity (95.0%) suitable for research-grade conjugation reactions.
  • White to off-white solid for easy handling.
  • Stable under recommended storage conditions to preserve activity.