Learn More
Medchemexpress LLC Azide-PEG-amine | 95.0% | 100 MG
SDP
Catalog No. 5000355466
Encompass_Preferred
Azide-PEG-amine (MW 5000) is a PEG-based PROTAC linker designed for the synthesis of PROTACs. This molecule functions as a versatile click chemistry reagent, enabling crucial reactions in chemical biology and drug discovery.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains an azide group
  • Participates in copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with alkyne groups
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups
Catalog No. 50-003-55466 Supplier Medchemexpress LLC Supplier No. HY140663100MG
Azide-PEG-amine (MW 5000) is a PEG-based PROTAC linker designed for the synthesis of PROTACs. This molecule functions as a versatile click chemistry reagent, enabling crucial reactions in chemical biology and drug discovery.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains an azide group
  • Participates in copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with alkyne groups
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups