Learn More
Medchemexpress LLC Azide-PEG-amine | 95.0% | 25 MG
SDP
Catalog No. 5000352625
Encompass_Preferred
Azide-PEG-amine (MW 2000) is a PEG-based PROTAC linker, essential for synthesizing PROTACs. It acts as a versatile click chemistry reagent, featuring an azide group. This enables its participation in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne groups, and strain-promoted alkyne-azide cycloaddition (SPAAC) with DBCO or BCN groups. PROTACs utilize this linker to connect ligands, targeting E3 ubiquitin ligase and a protein of interest for selective degradation. This product is for research use only.
  • PEG-based PROTAC linker for PROTAC synthesis
  • Functions as a click chemistry reagent with an azide group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Supports strain-promoted alkyne-azide cycloaddition (SPAAC)
  • Leverages ubiquitin-proteasome system for selective protein degradation
  • Intended for research use only
Catalog No. 50-003-52625 Supplier Medchemexpress LLC Supplier No. 25MGHY140661
Azide-PEG-amine (MW 2000) is a PEG-based PROTAC linker, essential for synthesizing PROTACs. It acts as a versatile click chemistry reagent, featuring an azide group. This enables its participation in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne groups, and strain-promoted alkyne-azide cycloaddition (SPAAC) with DBCO or BCN groups. PROTACs utilize this linker to connect ligands, targeting E3 ubiquitin ligase and a protein of interest for selective degradation. This product is for research use only.
  • PEG-based PROTAC linker for PROTAC synthesis
  • Functions as a click chemistry reagent with an azide group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Supports strain-promoted alkyne-azide cycloaddition (SPAAC)
  • Leverages ubiquitin-proteasome system for selective protein degradation
  • Intended for research use only