Learn More
Medchemexpress LLC Azide-PEG-amine (MW 2000) | 95.0% | 50 MG
SDP
Catalog No. 5000354593
Encompass_Preferred
Azide-PEG-amine (MW 2000) is a PEG-based PROTAC linker that functions as a click chemistry reagent. It contains an Azide group, enabling copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with Alkyne-containing molecules, and strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups. PROTACs, composed of two distinct ligands connected by a linker, leverage the intracellular ubiquitin-proteasome system for selective degradation of target proteins.
  • PEG-based PROTAC linker for PROTAC synthesis
  • Functions as a click chemistry reagent
  • Features an Azide group for CuAAc and SPAAC reactions
  • Average molecular weight of 2000
  • Appears as a white to off-white solid
  • Intended for research use only
Catalog No. 50-003-54593 Supplier Medchemexpress LLC Supplier No. HY14066150MG
Azide-PEG-amine (MW 2000) is a PEG-based PROTAC linker that functions as a click chemistry reagent. It contains an Azide group, enabling copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with Alkyne-containing molecules, and strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups. PROTACs, composed of two distinct ligands connected by a linker, leverage the intracellular ubiquitin-proteasome system for selective degradation of target proteins.
  • PEG-based PROTAC linker for PROTAC synthesis
  • Functions as a click chemistry reagent
  • Features an Azide group for CuAAc and SPAAC reactions
  • Average molecular weight of 2000
  • Appears as a white to off-white solid
  • Intended for research use only