Learn More
Medchemexpress LLC Azide-PEG-azide | 00-00-0 | 95.0% | 10MG
SDP
Catalog No. 5000260180
Encompass_Preferred
Azide-PEG-azide (MW 10000) is a bifunctional polyethylene glycol (PEG) linker with azide groups at both termini. It is designed for click chemistry conjugation and linker construction in chemical biology and medicinal chemistry, providing spacer length and solubility advantages for bioconjugation and PROTAC design.
  • High average molecular weight suitable for spacing and solubility optimization.
  • Azide termini enable copper-catalyzed azide-alkyne cycloaddition (click chemistry).
  • Good purity for research applications, supporting reproducible conjugation.
  • Available in small-scale and bulk quantities for screening and scale-up.
  • Stable under recommended storage, facilitating reliable long-term use.
Catalog No. 50-002-60180 Supplier Medchemexpress LLC Supplier No. HY14066610MG
Azide-PEG-azide (MW 10000) is a bifunctional polyethylene glycol (PEG) linker with azide groups at both termini. It is designed for click chemistry conjugation and linker construction in chemical biology and medicinal chemistry, providing spacer length and solubility advantages for bioconjugation and PROTAC design.
  • High average molecular weight suitable for spacing and solubility optimization.
  • Azide termini enable copper-catalyzed azide-alkyne cycloaddition (click chemistry).
  • Good purity for research applications, supporting reproducible conjugation.
  • Available in small-scale and bulk quantities for screening and scale-up.
  • Stable under recommended storage, facilitating reliable long-term use.