Learn More
Medchemexpress LLC Azide-PEG-azide | 95.0% | 10,000 Da (average) | 5 MG
SDP
Catalog No. 5000223184
Encompass_Preferred
Azide-PEG-azide is a bifunctional polyethylene glycol linker bearing azide groups at both termini, designed for use in click chemistry and bioconjugation workflows. The material is supplied as a polydisperse PEG with an average molecular weight of 10,000 Da and provides a hydrophilic, flexible spacer to connect molecular fragments, improving solubility and linker reach while enabling efficient azide-alkyne cycloaddition reactions.
  • Bifunctional PEG linker with terminal azide groups.
  • Average molecular weight 10,000 Da (polydisperse).
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAC) click chemistry.
  • Provides hydrophilic, flexible spacer to improve solubility and linker reach.
  • Suitable for bioconjugation, PEGylation, and linker synthesis applications.
Catalog No. 50-002-23184 Supplier Medchemexpress LLC Supplier No. HY1406665MG
Azide-PEG-azide is a bifunctional polyethylene glycol linker bearing azide groups at both termini, designed for use in click chemistry and bioconjugation workflows. The material is supplied as a polydisperse PEG with an average molecular weight of 10,000 Da and provides a hydrophilic, flexible spacer to connect molecular fragments, improving solubility and linker reach while enabling efficient azide-alkyne cycloaddition reactions.
  • Bifunctional PEG linker with terminal azide groups.
  • Average molecular weight 10,000 Da (polydisperse).
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAC) click chemistry.
  • Provides hydrophilic, flexible spacer to improve solubility and linker reach.
  • Suitable for bioconjugation, PEGylation, and linker synthesis applications.