Learn More
Medchemexpress LLC Azide-PEG-azide | 95.0% | 50 MG
SDP
Catalog No. 5000353513
Encompass_Preferred
Azide-PEG-azide is a PEG-based PROTAC linker used in the synthesis of PROTACs. It acts as a click chemistry reagent, featuring an Azide group that participates in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with alkyne-containing molecules. It also undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups. PROTACs utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains an azide group
  • Participates in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with alkyne groups
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups
  • Leverages the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-53513 Supplier Medchemexpress LLC Supplier No. HY14066650MG
Azide-PEG-azide is a PEG-based PROTAC linker used in the synthesis of PROTACs. It acts as a click chemistry reagent, featuring an Azide group that participates in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with alkyne-containing molecules. It also undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups. PROTACs utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains an azide group
  • Participates in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with alkyne groups
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups
  • Leverages the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins