Learn More
Medchemexpress LLC Azide-PEG12-alcohol | 1821464-55-4 | 99.8% | C24H49N3O12 | 250 MG
SDP
Catalog No. 5000352882
Encompass_Preferred
Azide-PEG12-alcohol is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. This compound functions as a click chemistry reagent, featuring an Azide group. It can engage in copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with molecules containing Alkyne groups. Additionally, it can participate in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules possessing DBCO or BCN groups.
  • PEG-based PROTAC linker
  • Click chemistry reagent with an azide group
  • Engages in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
  • Used in PROTAC synthesis
Catalog No. 50-003-52882 Supplier Medchemexpress LLC Supplier No. 250MGHY140803
Azide-PEG12-alcohol is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. This compound functions as a click chemistry reagent, featuring an Azide group. It can engage in copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with molecules containing Alkyne groups. Additionally, it can participate in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules possessing DBCO or BCN groups.
  • PEG-based PROTAC linker
  • Click chemistry reagent with an azide group
  • Engages in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
  • Used in PROTAC synthesis