Learn More
Medchemexpress LLC Azide-PEG12-alcohol | 1821464-55-4 | 99.8% | C24H49N3O12 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY14080350MG
Encompass_Preferred
Azide-PEG12-alcohol is a PEG-based PROTAC linker used in the synthesis of PROTACs. This click chemistry reagent contains an Azide group, enabling its use in various bioconjugation strategies.
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing alkyne groups.
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • Useful in PROTAC synthesis for targeted therapy research.
Catalog No. 50-003-53161