Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Azide-Peg3-Biotin Conjugate

Catalog No. 76202410MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
76-202-410MG 10 mg
76-202-425MG 25 mg
76-202-4100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 76-202-410MG Supplier Sigma Aldrich Supplier No. 76202410MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Biotinylation reagent for labeling alkyne containing molecules or biomolecules using either copper-catalyzed 1,3 dipolar cycloaddition click chemistry or copper-free click chemistry with cyclooctyne derivatives. The azide group reacts with alkynes to form a stable triazole linkage facilitating the introduction of biotin into your alkyne modified system of interest. This reaction is bioorthogonal or in other words compatible with biological systems in that it's components do not react with the biological environment.

Specifications

CAS 875770-34-6
Molecular Formula C18H32N6O5S
Linear Formula C18H32N6O5S
MDL Number MFCD20134145
Quantity 10 mg
Synonym N-[2-[2-[2-(2-Azidoethoxy)ethoxy]ethoxy]ethyl]hexahydro-2-oxo-(3aS,4S,6aR)- 1H-thieno[3,4-d ]imidazole-4-pentanamide; Biotin-PEG3-azide
Molecular Weight (g/mol) 444.55 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.