Learn More
Medchemexpress LLC Azide-PEG3-Desthiobi 10mg
SDP
Catalog No. 5000205723
Encompass_Preferred
Azide-PEG3-Desthiobiotin is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs[1] Azide-PEG3-Desthiobiotin is a click chemistry reagent it contains an Azide group and can undergo copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with molecules containing Alkyne groups It can also undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups
Catalog No. 50-002-05723 Supplier Medchemexpress LLC Supplier No. HYW09612010MG
Azide-PEG3-Desthiobiotin is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs[1] Azide-PEG3-Desthiobiotin is a click chemistry reagent it contains an Azide group and can undergo copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with molecules containing Alkyne groups It can also undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups