Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Azide-PEG3-tos | 178685-33-1 | 329.37 g/mol | C13H19N3O5S | 250 MG
SDP

Catalog No. 5000203937
Encompass_Preferred

Azide-PEG3-Tos is a heterobifunctional PEG linker containing an azide functional group for click chemistry. It acts as a non-cleavable, three-unit PEG linker used in the synthesis of PROTACs and antibody-drug conjugates, and participates in copper-catalyzed and strain-promoted azide-alkyne cycloaddition reactions.

  • Heterobifunctional PEG linker with azide group.
  • Molecular weight 329.37 g/mol.
  • Chemical formula C13H19N3O5S.
  • CAS number 178685-33-1.
  • Available in 250 MG packaging; other sizes available.
  • Storage: pure form -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Applications: PROTAC synthesis, ADC construction, click chemistry.

Catalog No. 50-002-03937 Supplier Medchemexpress LLC Supplier No. HY140004250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Azide-PEG3-Tos is a heterobifunctional PEG linker containing an azide functional group for click chemistry. It acts as a non-cleavable, three-unit PEG linker used in the synthesis of PROTACs and antibody-drug conjugates, and participates in copper-catalyzed and strain-promoted azide-alkyne cycloaddition reactions.

  • Heterobifunctional PEG linker with azide group.
  • Molecular weight 329.37 g/mol.
  • Chemical formula C13H19N3O5S.
  • CAS number 178685-33-1.
  • Available in 250 MG packaging; other sizes available.
  • Storage: pure form -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Applications: PROTAC synthesis, ADC construction, click chemistry.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.