Learn More
Medchemexpress LLC Azide-peg4-tos | 168640-82-2 | 99.6% | C15H23N3O6S | 100 MG
SDP
Catalog No. 5000353141
Encompass_Preferred
Azide-PEG4-Tos is a PEG-based PROTAC linker for PROTAC synthesis. It acts as a click chemistry reagent with an Azide group, enabling copper-catalyzed azide-alkyne cycloaddition (CuAAc) with Alkyne groups, or strain-promoted alkyne-azide cycloaddition (SPAAC) with DBCO or BCN groups. PROTACs use two linked ligands, one targeting an E3 ubiquitin ligase and the other a protein of interest, to selectively degrade proteins via the ubiquitin-proteasome system.
  • Peg-based PROTAC linker
  • Used in PROTAC synthesis
  • Click chemistry reagent with azide group
  • Undergoes CuAAc with alkyne groups
  • Participates in SPAAC with DBCO or BCN groups
  • PROTACs: two linked ligands
  • Targets E3 ubiquitin ligase and protein of interest
  • Utilizes ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-53141 Supplier Medchemexpress LLC Supplier No. HY140351100MG
Azide-PEG4-Tos is a PEG-based PROTAC linker for PROTAC synthesis. It acts as a click chemistry reagent with an Azide group, enabling copper-catalyzed azide-alkyne cycloaddition (CuAAc) with Alkyne groups, or strain-promoted alkyne-azide cycloaddition (SPAAC) with DBCO or BCN groups. PROTACs use two linked ligands, one targeting an E3 ubiquitin ligase and the other a protein of interest, to selectively degrade proteins via the ubiquitin-proteasome system.
  • Peg-based PROTAC linker
  • Used in PROTAC synthesis
  • Click chemistry reagent with azide group
  • Undergoes CuAAc with alkyne groups
  • Participates in SPAAC with DBCO or BCN groups
  • PROTACs: two linked ligands
  • Targets E3 ubiquitin ligase and protein of interest
  • Utilizes ubiquitin-proteasome system
  • Selectively degrades target proteins