Learn More
Medchemexpress LLC Azide-PEG4-Tos | 168640-82-2 | 99.6% | C15H23N3O6S | 500 MG
SDP
Catalog No. 5000357001
Encompass_Preferred
Azide-PEG4-Tos is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. This compound functions as a click chemistry reagent, featuring an Azide group. It can engage in copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with molecules containing Alkyne groups, as well as strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules possessing DBCO or BCN groups.
  • PEG-based PROTAC linker.
  • Click chemistry reagent.
  • Contains an azide group, enabling CuAAc reactions with alkyne groups.
  • Can participate in SPAAC reactions with DBCO or BCN groups.
Catalog No. 50-003-57001 Supplier Medchemexpress LLC Supplier No. HY140351500MG
Azide-PEG4-Tos is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. This compound functions as a click chemistry reagent, featuring an Azide group. It can engage in copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with molecules containing Alkyne groups, as well as strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules possessing DBCO or BCN groups.
  • PEG-based PROTAC linker.
  • Click chemistry reagent.
  • Contains an azide group, enabling CuAAc reactions with alkyne groups.
  • Can participate in SPAAC reactions with DBCO or BCN groups.