Learn More
Medchemexpress LLC Azide-PEG4-VC-PAB-doxorubicin | 99.6% | 1222.25 g·mol⁻1 | C57H75N9O21 | 50 MG
SDP
Catalog No. 5000204183
Encompass_Preferred
Azide-PEG4-VC-PAB-doxorubicin is an antibody-drug conjugate (ADC) agent-linker conjugate that couples the cytotoxic anthracycline doxorubicin to an azide-terminated PEG4-VC-PAB linker for use in bioconjugation and ADC assembly. It is designed for click chemistry conjugation (CuAAC and SPAAC) to alkyne- or DBCO/BCN-containing partners and is supplied as a solid for research use.
Catalog No. 50-002-04183 Supplier Medchemexpress LLC Supplier No. HY13628850MG
Azide-PEG4-VC-PAB-doxorubicin is an antibody-drug conjugate (ADC) agent-linker conjugate that couples the cytotoxic anthracycline doxorubicin to an azide-terminated PEG4-VC-PAB linker for use in bioconjugation and ADC assembly. It is designed for click chemistry conjugation (CuAAC and SPAAC) to alkyne- or DBCO/BCN-containing partners and is supplied as a solid for research use.