Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Azide-PEG4-VC-PAB-doxorubicin | 99.6% | 1222.25 g/mol | C57H75N9O21 | 10 MG
SDP

Catalog No. 5000204234
Encompass_Preferred

Azide-PEG4-VC-PAB-doxorubicin is a drug-linker conjugate used in antibody-drug conjugate research and click-chemistry bioconjugation.

  • Contains an azide functional group for click chemistry.
  • Valine-citrulline peptide linker with a PAB self-immolative spacer.
  • Molecular weight 1222.25 g/mol.
  • Chemical formula C57H75N9O21.
  • Purity 99.6%.
  • Pack size includes 10 mg solid and other sizes.
  • Appearance orange to red solid.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-04234 Supplier Medchemexpress LLC Supplier No. HY13628810MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Azide-PEG4-VC-PAB-doxorubicin is a drug-linker conjugate used in antibody-drug conjugate research and click-chemistry bioconjugation.

  • Contains an azide functional group for click chemistry.
  • Valine-citrulline peptide linker with a PAB self-immolative spacer.
  • Molecular weight 1222.25 g/mol.
  • Chemical formula C57H75N9O21.
  • Purity 99.6%.
  • Pack size includes 10 mg solid and other sizes.
  • Appearance orange to red solid.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.