Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Azide-PEG4-VC-PAB-doxorubicin | 99.6% | 1222.25 g/mol | C57H75N9O21 | 100 MG
SDP

Catalog No. 5000204233
Encompass_Preferred

Azide-PEG4-VC-PAB-doxorubicin is an azide-functionalized drug-linker conjugate that links the cytotoxic agent doxorubicin to a PEG4-VC-PAB linker for use in antibody-drug conjugate synthesis and click chemistry applications.

  • High purity (99.62%).
  • Molecular weight 1222.25 g/mol.
  • Chemical formula C57H75N9O21.
  • Orange to red solid appearance.
  • Storage: powder at -20°C (long term) or 4°C (short term); in solvent at -80°C or -20°C as recommended.
  • Available as solid (1-100 mg) and 10 mM solution in DMSO.

Catalog No. 50-002-04233 Supplier Medchemexpress LLC Supplier No. HY136288100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Azide-PEG4-VC-PAB-doxorubicin is an azide-functionalized drug-linker conjugate that links the cytotoxic agent doxorubicin to a PEG4-VC-PAB linker for use in antibody-drug conjugate synthesis and click chemistry applications.

  • High purity (99.62%).
  • Molecular weight 1222.25 g/mol.
  • Chemical formula C57H75N9O21.
  • Orange to red solid appearance.
  • Storage: powder at -20°C (long term) or 4°C (short term); in solvent at -80°C or -20°C as recommended.
  • Available as solid (1-100 mg) and 10 mM solution in DMSO.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.