Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Azide-PEG4-VC-PAB-doxorubicin | 99.6% | 1222.25 g/mol | C57H75N9O21 | 5 MG
SDP

Catalog No. 5000202598
Encompass_Preferred

Azide-PEG4-VC-PAB-doxorubicin is a drug-linker conjugate that links the cytotoxic agent doxorubicin to an azide-PEG4-VC-PAB cleavable linker, intended for use in antibody-drug conjugate (ADC) construction and click-chemistry conjugation (CuAAC and SPAAC).

  • Contains an azide functional group for click conjugation.
  • Compatible with copper-catalyzed azide-alkyne cycloaddition and strain-promoted alkyne-azide cycloaddition reactions.
  • Features a cleavable valine-citrulline-p-aminobenzyl (VC-PAB) linker for intracellular release.
  • Supplied as an orange to red solid for convenient handling and storage.
  • Purity 99.6% and molecular weight 1222.25 g/mol.
  • Available in small-mass pack sizes suitable for research-scale ADC synthesis.

Catalog No. 50-002-02598 Supplier Medchemexpress LLC Supplier No. HY1362885MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Azide-PEG4-VC-PAB-doxorubicin is a drug-linker conjugate that links the cytotoxic agent doxorubicin to an azide-PEG4-VC-PAB cleavable linker, intended for use in antibody-drug conjugate (ADC) construction and click-chemistry conjugation (CuAAC and SPAAC).

  • Contains an azide functional group for click conjugation.
  • Compatible with copper-catalyzed azide-alkyne cycloaddition and strain-promoted alkyne-azide cycloaddition reactions.
  • Features a cleavable valine-citrulline-p-aminobenzyl (VC-PAB) linker for intracellular release.
  • Supplied as an orange to red solid for convenient handling and storage.
  • Purity 99.6% and molecular weight 1222.25 g/mol.
  • Available in small-mass pack sizes suitable for research-scale ADC synthesis.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.