Learn More
Medchemexpress LLC Azide-PEG5-Boc | 1415800-41-7 | C17H33N3O7 | 100 MG
SDP
Catalog No. 5000356655
Encompass_Preferred
Azide-PEG5-Boc is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It is a click chemistry reagent containing an Azide group, capable of undergoing copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with molecules containing Alkyne groups. It can also undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • PEG-based PROTAC linker
  • Can be used in the synthesis of PROTACs
  • Click chemistry reagent
  • Contains an Azide group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc)
  • Compatible with molecules containing Alkyne groups
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
  • Compatible with molecules containing DBCO or BCN groups
Catalog No. 50-003-56655 Supplier Medchemexpress LLC Supplier No. HY140778100MG
Azide-PEG5-Boc is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It is a click chemistry reagent containing an Azide group, capable of undergoing copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with molecules containing Alkyne groups. It can also undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • PEG-based PROTAC linker
  • Can be used in the synthesis of PROTACs
  • Click chemistry reagent
  • Contains an Azide group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc)
  • Compatible with molecules containing Alkyne groups
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
  • Compatible with molecules containing DBCO or BCN groups