Learn More
Medchemexpress LLC Azide-PEG5-Boc | 1415800-41-7 | C17H33N3O7 | 500 MG
SDP
Catalog No. 5000353796
Encompass_Preferred
Azide-PEG5-Boc is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent, featuring an Azide group that enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne groups, and strain-promoted alkyne-azide cycloaddition (SPAAC) with DBCO or BCN groups. PROTACs are designed to selectively degrade target proteins by utilizing the intracellular ubiquitin-proteasome system.
  • Used in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains an Azide group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Enables strain-promoted alkyne-azide cycloaddition (SPAAC)
  • Utilizes the intracellular ubiquitin-proteasome system to degrade target proteins
  • For research use only
Catalog No. 50-003-53796 Supplier Medchemexpress LLC Supplier No. HY140778500MG
Azide-PEG5-Boc is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent, featuring an Azide group that enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne groups, and strain-promoted alkyne-azide cycloaddition (SPAAC) with DBCO or BCN groups. PROTACs are designed to selectively degrade target proteins by utilizing the intracellular ubiquitin-proteasome system.
  • Used in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains an Azide group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Enables strain-promoted alkyne-azide cycloaddition (SPAAC)
  • Utilizes the intracellular ubiquitin-proteasome system to degrade target proteins
  • For research use only