Learn More
Medchemexpress LLC Azide-PEG5-tos | 236754-49-7 | 98.5% | 417.48 g·mol⁻¹ | C17H27N3O7S | 1 G
SDP
Catalog No. 5000202678
Encompass_Preferred
Azide-PEG5-Tos is a cleavable 5-unit polyethylene glycol (PEG) linker bearing an azide functional group and a tosyl terminus, designed for bioconjugation and ADC linker synthesis. The PEG spacer increases aqueous solubility and the azide enables click chemistry reactions such as CuAAC and SPAAC.
  • Cleavable 5-unit PEG spacer increases aqueous solubility.
  • Azide functional group enables CuAAC and SPAAC click reactions.
  • Tosyl terminus acts as a leaving group for nucleophilic substitution.
  • Available in multiple pack sizes, including 1 G.
  • High purity (98.5%) and characterized molecular weight 417.48 g·mol⁻¹.
Catalog No. 50-002-02678 Supplier Medchemexpress LLC Supplier No. HY1403521G
Azide-PEG5-Tos is a cleavable 5-unit polyethylene glycol (PEG) linker bearing an azide functional group and a tosyl terminus, designed for bioconjugation and ADC linker synthesis. The PEG spacer increases aqueous solubility and the azide enables click chemistry reactions such as CuAAC and SPAAC.
  • Cleavable 5-unit PEG spacer increases aqueous solubility.
  • Azide functional group enables CuAAC and SPAAC click reactions.
  • Tosyl terminus acts as a leaving group for nucleophilic substitution.
  • Available in multiple pack sizes, including 1 G.
  • High purity (98.5%) and characterized molecular weight 417.48 g·mol⁻¹.